Learn More
Novus Biologicals™ Human Caspase-8 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP183733
Catalog No. NBP183733
Add to cart
Description
Description
The Human Caspase-8 ELISA Kit (Colorimetric) measures human Caspase-8 in cell culture supernatant, cell lysate, serum.
Specifications
Specifications
0.16 - 10.0ng/mL
841
ELISA Kit
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cancer, Caspases, Cell Biology, Ubiquitin Proteasome Pathway
AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Human
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Caspase-8, anti-human Caspase-8 polyclonal (rabbit) Detection Antibody, Anti-rabbit-IgG-HRP, Human Caspase-8 Standard lyophilized, 20.00ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Lysis Buffer 10x, Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
0.10ng/mL
Caspase-8
CASP8
96 Tests
Cell Culture Supernatant, Cell Lysates, Serum