Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cathepsin B Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 17 publications
Supplier: R&D Systems AF953SP
This item is not returnable.
View return policy
Description
Cathepsin B Polyclonal specifically detects Cathepsin B in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Knockout Validated.Specifications
Cathepsin B | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, CTSB, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
Goat | |
Affinity Purified | |
RUO | |
1508 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunoassay | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-25 ug/mL, Knockout Validated | |
P07858 | |
CTSB | |
Mouse myeloma cell line NS0-derived recombinant human Cathepsin B Arg18-Ile339 Accession # P07858 | |
25 μg | |
Primary | |
Detects human Cathepsin B in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant mouse (rm) Cathepsin B is observed and less than 5% cross-reactivity with recombinant human (rh) Cathepsin C, rmCathepsin H, and rhCathepsin L is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction