Learn More
Novus Biologicals™ Human Cathepsin D ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP267257
Description
The Human Cathepsin D ELISA Kit (Colorimetric) measures human Cathepsin D in serum, plasma and other biological fluids.Specifications
46.88 - 3000 pg/mL | |
This kit recognizes Human CTSD in samples. No significant cross-reactivity or interference between Human CTSD and analogues was observed. | |
1509 | |
Human | |
1 Kit | |
Storage is content dependent. |
28.13 pg/mL | |
ELISA | |
CTSD | |
ELISA Kit (Colorimetric) | |
Biological Fluids,Plasma,Serum | |
Cathepsin D |