Learn More
Novus Biologicals™ Human Cathepsin L ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191259
Catalog No. NBP191259
Add to cart
Description
Description
The Human Cathepsin L ELISA Kit (Colorimetric) measures human Cathepsin L in cell culture supernatant, serum.
Specifications
Specifications
3.1 - 50ng/mL
1514
ELISA Kit
1.7ng/mL
Human (Hu)
Cathepsin L
CTSL1
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cathepsin L, Biotin-Conjugate anti-human Cathepsin L monoclonal antibody, Streptavidin-HRP, Human Cathepsin L Standard lyophilized, 100ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Cancer, Cellular Markers
cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP