Learn More
Novus Biologicals™ Human CCL1/I-309/TCA-3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275063
Catalog No. NBP275063
Add to cart
Description
Description
The Human CCL1/I-309/TCA-3 ELISA Kit (Colorimetric) measures human CCL1/I-309/TCA-3 in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 - 2000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CCL1 in samples. No significant cross-reactivity or interference between Human CCL1 and analogues was observed.
6346
18.75 pg/mL
CCL1/I-309/TCA-3
CCL1
Biological Fluids, Plasma, Serum