Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CCL11/Eotaxin Luminex Performance Assay

Bead-based multiplex assay for the Luminex™ platform

Manufacturer:  R&D Systems™ LUB320

Catalog No. LUB320

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

  • User selects specific analytes of interest
  • Requires small sample size
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
Specifications

Specifications

8.2pg/mL
Polystyrene
16.00 - 12000pg/mL
6356
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Cell Culture, Plasma (EDTA, Heparin), Serum
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Human
1 Kit
CCL11/Eotaxin
CCL11
Bead-based Multiplex Assay for the Luminex™ Platform
Cell Culture Supernates (50μL),Serum (15μL),EDTA Plasma (15μL),Heparin Plasma (15μL)
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit