Learn More
Novus Biologicals™ Human CCL2/MCP1 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191276
Catalog No. NBP191276
Add to cart
Description
Description
The Human CCL2/MCP1 ELISA Kit (Colorimetric) measures human CCL2/MCP1 in cell culture supernatant, serum, plasma (edta, heparin).
Specifications
Specifications
15.6 - 1,000pg/mL
6347
ELISA Kit
2.3pg/mL
Human (Hu)
CCL2/MCP1
CCL2
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MCP-1, HRP-Conjugate anti-human MCP-1 monoclonal antibody, Human MCP-1 Standard lyophilized, 2ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, heparin)
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant pro