Learn More
Gibco™ Human CCL3 (MIP-1 alpha) Recombinant Protein, PeproTech®

E. coli Recombinant Protein
$87.00 - $3240.00
Specifications
Accession Number | P10147 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 6348 |
Molecular Weight (g/mol) | 7.8 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
30008100UG
![]() ![]() |
Gibco™
30008100UG |
100 µg |
Each for $648.00
|
|
|||||
300-08-1MG
![]() ![]() |
Gibco™
300081MG |
1 mg |
Each for $3,240.00
|
|
|||||
300-08-20UG
![]() ![]() |
Gibco™
3000820UG |
20 µg |
Each for $211.00
|
|
|||||
30008250UG
![]() ![]() |
Gibco™
30008250UG |
250 µg |
Each for $1,134.00
|
|
|||||
30008500UG
![]() ![]() |
Gibco™
30008500UG |
500 µg |
Each for $1,944.00
|
|
|||||
300-08-50UG
![]() ![]() |
Gibco™
3000850UG |
50 µg |
Each for $389.00
|
|
|||||
300-08-5UG
![]() ![]() |
Gibco™
300085UG |
5 µg |
Each for $87.00
|
|
|||||
Description
Both MIP-1alpha and MIP-1beta are structurally and functionally related CC chemokines. They participate in host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells (e.g. macrophages, lymphocytes and NK cells). While both MIP-1alpha and MIP-1beta exert similar effects on monocytes, their effect on lymphocytes differ; with MIP-1alpha selectively attracting CD8+ lymphocytes, and MIP-1beta selectively attracting CD4+ lymphocytes. Additionally, MIP-1alpha and MIP-1beta have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1alpha and MIP-1beta are active on human and murine hematopoietic cells.Specifications
P10147 | |
protein with no preservative | |
7.8 | |
E. coli | |
RUO | |
AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; CCL3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78ALPHA; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1 alpha; MIP1 (a); mip1 alpha; MIP-1 alpha; MIP1-(a); MIP1A; MIP-1 A; MIP-1 alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; Scya3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1 A); small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
6348 | |
Human CCL3 (MIP-1 alpha) | |
-20°C | |
<1 EU/µg | |
Ccl3 | |
Unconjugated | |
ASLAADTPTA CCFSYTSRQI PQNFIADYFE TSSQCSKPGV IFLTKRSRQV CADPSEEWVQ KYVSDLELSA | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.