Learn More
Invitrogen™ eBioscience™ Human CCL4 (MIP-1 beta) ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.
Specifically engineered for accurate and precise measurement of CCL4 protein levels from samples including serum, plasma, and cell culture supernatants.
Manufacturer:  Invitrogen™ 88703476
 View more versions of this product
Catalog No. 50-112-5176
Add to cart
Alert:
Contains Sodium azide
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution
Description
Description
CCL4 (macrophage inflammatory protein 1-beta, MIP1B) belongs to the intercrine beta (chemokine CC) family. Functionally, CCL4 is involved in chemotactic and proinflammatory effects, and homeostasis. CCL4 is produced by macrophages upon stimulation by bacterial endotoxins. CCL4 recruits and stimulates various inflammatory cells at sites of inflammation. CCL4 is produced by lymphocytes, macrophages and dendritic cells. Both CCL4 and the related protein CCL3 participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells. While both CCL4 and CCL3 exert similar effects on monocytes, their effect on lymphocytes differ, with CCL4 selectively attracting CD4+ lymphocytes and CCL3 selectively attracting CD8+ lymphocytes. Additionally, both have been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. The processed form of CCL4 can induce down-modulation of surface expression of the chemokine receptor CCR5, thus inhibiting the CCR5-mediated entry of HIV-1 in T cells. CCL4 binds with high affinity to CCR5 receptors.
Specifications
Specifications
P13236
Biotin
6351
Colorimetric Microplate Reader
ELISA Kit
RUO
Plasma,Serum,Supernatant
Human
1 hr 45 min
Human
C-C motif Chemokine 4, Macrophage Inflammatory Protein 1 beta
4-500 pg/mL
ELISA
96-well plates
10X Coating Buffer: Buffer for plating the Capture Antibody
20 x 96 tests
4 pg/mL
Plasma,100 μL|Serum,100 μL|Supernatant,100 μL
2-8°C
24 hr 30 min
CCL4
HRP