Learn More
Novus Biologicals™ Human CCL4L1/LAG-1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275067
Description
The Human CCL4L1/LAG-1 ELISA Kit (Colorimetric) measures human CCL4L1/LAG-1 in serum, plasma and other biological fluids.Specifications
15.63 - 1000 pg/mL | |
This kit recognizes Human CCL4L1 in samples. No significant cross-reactivity or interference between Human CCL4L1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
9560 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
9.38 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CCL4L2 | |
1 Kit | |
CCL4L1/LAG-1 |