Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCL5 (RANTES) (aa 24-91), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$160.50 - $2060.00

Specifications

Recombinant Yes
Name Human CCL5 (RANTES) (aa 24-91)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87853 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87853
10 µg
Each for $160.50
Add to cart
 
PIRP87854 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87854
25 µg
Each for $289.00
Add to cart
 
PIRP87855 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87855
100 µg
Each for $770.50
Add to cart
 
PIRP87856 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87856
500 µg
Each for $2,060.00
Add to cart
 
Description

Description

RANTES is a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. It binds to CCR1, CCR3, CCR4 and CCR5 and is one of the major HIV suppressive factors produced by CD8+ T cells. Recombinant RANTES protein induces a dose dependent inhibition of different strains of HIV 1, HIV 2, and simian immunodeficiency virus (SIV). It is T cell and macrophage specific, induced by mitogens, and belongs to the intercrine beta (chemokine CC) family.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
0.1% TFA with 26% acetonitrile and no preservative
E. coli
-20°C
P13501
CCL5
>95%
Human CCL5 (RANTES) (aa 24-91)
200 µg/mL
Liquid
beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; eoCP; Eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; SCYA5; SIS d; SISd; SIS-delta; SISd; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014)
RUO
Human
6352
7.9 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit