Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCP-2/CCL8 Human Uncoated ELISA Kit with Plates Non-distribution product as customer accommodation.

Uncoated ELISA Kit with Plates

Manufacturer:  Invitrogen™ 885216922

Catalog No. 50-112-5170


Add to cart

Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Human CCL8 (MCP-2) Standard (lyophilized): 20pg/mL upon reconstitution
Coating Buffer Concentrate
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent Y
pre-titrated Streptavidin-HRP
Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

The Human Monocyte Chemoattractant Protein 2 (CCL8) MCP-2) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human MCP-2. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
Specifications

Specifications

P80075
Biotin
6355
Colorimetric Microplate Reader
ELISA Kit
RUO
Plasma,Serum,Supernatant
Plasma,10 μL|Serum,10 μL|Supernatant,10 μL
2-8°C
24 hr 30 min
CCL8
HRP
0.16-10 pg/mL
ELISA
96-well plates
10X Coating Buffer: Buffer for plating the Capture Antibody
2 x 96 tests
0.16 pg/mL
10μL
Human
1 hr 45 min
Human
Monocyte chemoattractant protein 2
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit