Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CCNDBP1 Full-length ORF (AAH09689.1, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name CCNDBP1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 66.7kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-938-499
View Documents
Abnova™
H00023582P01L
25 ug
Each for $709.50
Add to cart
 
89-938-498
View Documents
Abnova™
H00023582P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene was identified by the interaction of its gene product with Grap2, a leukocyte-specific adaptor protein important for immune cell signaling. The protein encoded by this gene was shown to interact with cyclin D. Transfection of this gene in cells was reported to reduce the phosphorylation of Rb gene product by cyclin D-dependent protein kinase, and inhibit E2F1-mediated transcription activity. This protein was also found to interact with helix-loop-helix protein E12 and is thought to be a negative regulator of liver-specific gene expression. Several alternatively spliced variants have been found for this gene. [provided by RefSeq]

Sequence: MASATAPAAAVPTLASPLEQLRHLAEELRLLLPRVRVGEAQETTEEFNREMFWRRLNEAAVTVSREATTLTIVFSQLPLPSPQETQKFCEQVHAAIKAFIAVYYLLPKDQGITLRKLVRGATLDIVDGMAQLMEVLSVTPTQSPENNDLISYNSVWVACQQMPQIPRDNKAAALLMLTKNVDFVKDAHEEMEQAVEECDPYSGLLNDTEENNSDNHNHEDDVLGFPSNQDLYWSEDDQELIIPCLALVRASKACLKKIRMLVAENGKKDQVAQLDDIVDISDEISPSVDDLALSIYPPMCHLTVRINSAKLVSVLKKALEITKASHVTPQPEDSWIPLLINAIDHCMNRIKELTQSELEL
Specifications

Specifications

CCNDBP1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
66.7kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit