Learn More
Novus Biologicals™ Human CCR11 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275119
Description
The Human CCR11 ELISA Kit (Colorimetric) measures human CCR11 in serum, plasma and other biological fluids.Specifications
1.56 - 100 ng/mL | |
This kit recognizes Human CCRL1 in samples. No significant cross-reactivity or interference between Human CCRL1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
51554 | |
Human | |
GPCR | |
CCR11 |
0.94 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
ACKR4 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |