Learn More
ACROBiosystems ACROBiosystems Human CCR8 Full Length Protein-VLP (HEK293)
Supplier:  ACROBiosystems CC8H52P420UG
Encompass_Preferred
Recombinant Protein;20UG;Human CCR8 Full Length Protein-VLP (CC8-H52P4) is expressed from human 293 cells (HEK293). It contains AA Asp 2 - Leu 355 (Accession # P51685-1).;This protein carries no "tag". The protein has a calculated MW of 40.7 kDa.
Catalog No. 50-210-6961