Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human CD142 Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
$89.50 - $5770.00
Specifications
Accession Number | P13726 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2152 |
Molecular Weight (g/mol) | 51.6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
15019T100UG
|
Gibco™
15019T100UG |
100 μg |
Each for $1,154.00
|
|
|||||
15019T10UG
|
Gibco™
15019T10UG |
10 μg |
Each for $217.00
|
|
|||||
150-19T-1MG
|
Gibco™
15019T1MG |
1 mg |
Each for $5,770.00
|
|
|||||
15019T250UG
|
Gibco™
15019T250UG |
250 μg |
Each for $2,020.00
|
|
|||||
150-19T-2UG
|
Gibco™
15019T2UG |
2 μg |
Each for $89.50
|
|
|||||
15019T500UG
|
Gibco™
15019T500UG |
500 μg |
Each for $3,460.00
|
|
|||||
15019T50UG
|
Gibco™
15019T50UG |
50 μg |
Each for $692.00
|
|
|||||
Description
This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described.Specifications
P13726 | |
protein with no preservative | |
51.6 | |
CHO cells | |
RUO | |
AA409063; CD142; Cf3; Cf-3; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); coagulation factor III, tissue factor; F3; TF; TFA; Thromboplastin; Tissue factor | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
2152 | |
Human CD142 | |
-20°C | |
<1 EU/μg | |
F3 | |
Unconjugated | |
SGTTNTVAAY NLTWKSTNFK TILEWEPKPV NQVYTVQIST KSGDWKSKCF YTTDTECDLT DEIVKDVKQT YLARVFSYPA GNVESTGSAG EPLYENSPEF TPYLETNLGQ PTIQSFEQVG TKVNVTVEDE RTLVRRNNTF LSLRDVFGKD LIYTLYYWKS SSSGKKTAKT NTNEFLIDVD KGENYCFSVQ AVIPSRTVNR KSTDSPVECM GGEKGEFREH HHHHHHH | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title