Learn More
Novus Biologicals™ Human CD23/Fc epsilon RII ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191173
Catalog No. NBP191173
Add to cart
Description
Description
The Human CD23/Fc epsilon RII ELISA Kit (Colorimetric) measures human CD23/Fc epsilon RII in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
12.5 - 400 U/mL
2208
ELISA Kit
6.8 U/mL
Human (Hu)
CD23/Fc epsilon RII
FCER2
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD23, Biotin-Conjugate anti-human sCD23 monoclonal antibody, Streptavidin-HRP, Human sCD23 Standard lyophilized, 400U/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Cancer, Immunology
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RI