Learn More
ACROBiosystems ACROBiosystems Human CD24 Protein, His tag (Nanoparticle)
Supplier:  ACROBiosystems CD4H52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD24, His tag (Nanoparticle) (CD4-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession # P25063-1).;Human CD24 Protein, His tag (Nanoparticle) is designed and expressed as nanoparticles of approximately 25 nm diameter by displaying CD24 ECD in native conformation. The nanoparticles each was coated with an average of 180 CD24 ECD.
The nanoparticle technology allows directional and high-density display of human CD24 on the surface of nanoparticles, thus greatly enhances the immune response in vivo to otherwise poorly immunogenic epitopes.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.3 kDa. The protein migrates as 43-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD24, His tag (Nanoparticle) (Cat. No. CD4-H52H5) at 5 μg/mL (100 μL/well) can bind Anti-CD24 MAb (SN3) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-210-6337
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.