Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD25 Carrier-Free, Recombinant Protein, eBioscience™ Non-distribution product as customer accommodation.

Recombinant Protein

Manufacturer:  Invitrogen 34870282

 View more versions of this product

Catalog No. 50-112-3523


Add to cart

Description

Description

CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency. Description: The human soluble IL-2 receptor alpha chain (sIL-2Ra) is generated by proteolytic cleavage of the cell-surface bound receptor on activated T cells. The IL-2Ra (also known as CD25) is expressed by early progenitors of the T and B lineage, as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 with low affinity. However, association with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) results in formation of the high affinity IL-2R. CD25 plays a role in lymphocyte differentiation and activation/proliferation. sIL-2Ra has been detected in the serum of patients with autoimmune disease and leukemia. Reports have demonstrated that addition of sIL-2Ra can inhibit IL-2-mediated signaling and enhance Th17 responses. Applications Reported: Recombinant human soluble IL-2Ra is biologically active and can enhance proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2. Applications Tested: The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2, is 0.5-1.5ug/mL. Source: Insect cells (amino acids Glu22-Gln240; accession &Num;: NP_000408.1). Bioactivity: The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2, is 0.5-1.5ug/mL. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE and HPLC. Molecular Weight: 24.8 kDa. Storage and handling: For best recovery, quick spin vial prior to opening. Reconstitute in 1X PBS, pH 7.2 to a concentration of 0.1-1.0mg/mL. Do not vortex. Use in sterile environment. Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier-protein (e.g., 0.1% BSA) at less than or equal to -20°C.
Specifications

Specifications

P01589
Lyophilized
3559
Human CD25 Carrier-Free
Affinity chromatography
Insect Cells
RUO
Recombinant
Human CD25 Carrier-Free
Control, Functional Assay
Recombinant Protein, Carrier-Free
Unlabeled
Insect cells
Control, Functional Assay
PBS with no preservative
24.8kDa
SDS-PAGE and HPLC
100μg
-20°C
Insect cells (amino acids Glu22- Gln240; Accession #: NP_000408.1)
Less than 0.1ng/μg cytokine as determined by the LAL assay
IL2RA
The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2, is 0.5-1.5μg/mL.
Human
Hu
Recombinant
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit