Learn More
Novus Biologicals™ Human CD3 gamma ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275144
Catalog No. NBP275144
Add to cart
Description
Description
The Human CD3 gamma ELISA Kit (Colorimetric) measures human CD3 gamma in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CD3g in samples. No significant cross-reactivity or interference between Human CD3g and analogues was observed.
917
0.10 ng/mL
CD3 gamma
CD3G
Biological Fluids, Plasma, Serum