Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ Human CD30/TNFRSF8 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191185
Catalog No. NBP191185
Description
Description
The Human CD30/TNFRSF8 ELISA Kit (Colorimetric) measures human CD30/TNFRSF8 in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
1.6 - 100ng/mL
943
ELISA Kit
0.3ng/mL
Human (Hu)
CD30/TNFRSF8
TNFRSF8
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD30, HRP-Conjugate anti-human sCD30 monoclonal antibody, Human sCD30 Standard lyophilized, 200ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cell Cycle and Replication, Embryonic Stem Cell Markers, Immunology, Stem Cell Markers
CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8