Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD38 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF2404
This item is not returnable.
View return policy
Description
CD38 Polyclonal specifically detects CD38 in Human samples. It is validated for Western Blot.Specifications
CD38 | |
Polyclonal | |
Unconjugated | |
P28907 | |
CD38 | |
Mouse myeloma cell line NS0-derived recombinant human CD38 Val43-Ile300 Accession # P28907 | |
100 μg | |
Primary | |
Detects human CD38 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse CD38 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 0.1-15 ug/mL | |
2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl Cyclase, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose Hydrolase, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
952 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction