Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human CD40 Ligand/TNFSF5 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP191184
Description
The Human CD40 Ligand/TNFSF5 ELISA Kit (Colorimetric) measures human CD40 Ligand/TNFSF5 in cell culture supernatant, serum, plasma (edta, citrate, heparin).Specifications
0.16 - 10.0ng/mL | |
ELISA Kit | |
The assay detects both natural and recombinant human CD40 Ligand/TNFSF5. The cross reactivity and interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human CD40 Ligand/TNFSF5 positive sample. There was no cross reactivity or interference. | |
ELISA | |
CD40LG | |
96 Tests | |
Storage of components varies. See protocol for specific instructions. | |
CD40 Ligand/TNFSF5 |
0.06ng/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
Human | |
959 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD40L, HRP-Conjugate anti-human sCD40L monoclonal antibody, Human sCD40L Standard lyophilized, 20ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films, Dilution Plates | |
Adaptive Immunity, Apoptosis, Asthma, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Ma | |
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 ( |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction