Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit
Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$736.60 - $4199.20
Specifications
Assay Range | 62.50 to 4000 pg/mL |
---|---|
Assay Sensitivity | 10.1 pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Product Type | Solid Phase Sandwich ELISA |
Catalog Number | Mfr. No. | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Sufficient For | Price | Quantity & Availability | ||||
DCDL40
|
R&D Systems™
DCDL40 |
1 Plate | Sufficient materials to run ELISAs on at least one 96-well plate |
Each for $736.60
|
|
||||
SCDL40
|
R&D Systems™
SCDL40 |
1 Pk. (6 Plates) | Sufficient materials to run ELISAs on at least six 96-well plates |
Each for $4,199.20
|
|
||||
Description
The Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CD40 Ligand/TNFSF5 in cell culture supernates (100 ul), serum (20 ul), platelet-poor edta plasma (100 ul), platelet-poor heparin plasma (100 ul).Specifications
62.50 to 4000 pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human CD40 Ligand | |
ELISA | |
CD40LG | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
CD40 Ligand/TNFSF5 |
10.1 pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Heparin), Serum | |
Human | |
959 | |
Cell Culture Supernates (100uL),Serum (20uL),Platelet-poor EDTA Plasma (100uL),Platelet-poor Heparin Plasma (100uL) | |
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40L, CD40-L, CD40LG, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5, TNFSF5IMD3, TRAP, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title