Learn More
Novus Biologicals™ Human CD40/TNFRSF5 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191264
Catalog No. NBP191264
Add to cart
Description
Description
The Human CD40/TNFRSF5 ELISA Kit (Colorimetric) measures human CD40/TNFRSF5 in cell culture supernatant, serum, plasma (edta, heparin), amniotic fluid.
Specifications
Specifications
7.8 - 500pg/mL
958
ELISA Kit
1.3pg/mL
Human (Hu)
CD40/TNFRSF5
CD40
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD40, Biotin-Conjugate anti-human sCD40 monoclonal antibody, Streptavidin-HRP, Human sCD40 Standard lyophilized, 1ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, heparin), amniotic fluid
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Apoptosis, Asthma, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocy