Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CD47 His-tag Recombinant Protein, Carrier-Free Available on GSA/VA Contract for Federal Government customers only.

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Manufacturer:  R&D Systems™ 9396CD100

Catalog No. 93-96C-D100


Add to cart
This item is not returnable. View return policy

Description

Description

The Recombinant Human CD47 His-tag Protein is derived from NS0. The Recombinant Human CD47 His-tag Protein has been validated for the following applications: Bioactivity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Ships at ambient temperature.

Specifications

Specifications

Q08722
Lyophilized from a 0.2μm filtered solution in PBS
15 kDa
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.10 EU per 1μg of the protein by the LAL method.
0.04 to 0.2μg/mL
Unlabeled
Yes
Bioactivity
961
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Mouse myeloma cell line, NS0-derived Gln19-Pro139, with a C-terminal 6-His tag
RUO
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen
Recombinant protein
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.