Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CD93 Partial ORF (ABM85160.1, 24 a.a. - 580 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name CD93 (Human) Recombinant Protein (Q03)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 86.9kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-938-804
View Documents
Abnova™
H00022918Q03L
25 ug
Each for $709.50
Add to cart
 
89-938-803
View Documents
Abnova™
H00022918Q03S
10 ug
Each for $460.50
Add to cart
 
Description

Description

The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton. [provided by RefSeq]

Sequence: ADTEAVVCVGTACYTAHSGKLSAAEAQNHCNQNGGNLATVKSKEEAQHVQRVLAQLLRREAALTARMSKFWIGLQREKGKCLDPSLPLKGFSWVGGGEDTPYSNWHKELRNSCISKRCVSLLLDLSQPLLPSRLPKWSEGPCGSPGSPGSNIEGFVCKFSFKGMCRPLALGGPGQVTYTTPFQTTSSSLEAVPFASAANVACGEGDKDETQSHYFLCKEKAPDVFDWGSSGPLCVSPKYGCNFNNGGCHQDCFEGGDGSFLCGCRPGFRLLDDLVTCASRNPCSSSPCRGGATCVLGPHGKNYTCRCPQGYQLDSSQLDCVDVDECQDSPCAQECVNTPGGFRCECWVGYEPGGPGEGACQDVDECALGRSPCAQGCTNTDGSFHCSCEEGYVLAGEDGTQCQDVDECVGPGGPLCDSLCFNTQGSFHCGCLPGWVLAPNGVSCTMGPVSLGPPSGPPDEEDKGEKEGSTVPRAATASPTRGPEGTPKATPTTSRPSLSSDAPITSAPLKMLAPSGSPGVWREPSIHHATAASGPQEPAGGDSSVATQNNDGTDGQK
Specifications

Specifications

CD93 (Human) Recombinant Protein (Q03)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
86.9kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit