Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD96 His-tag, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$770.50

Specifications

Recombinant Yes
Name SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
For Use With (Application) Control, ELISA, Western Blot
Conjugate Unlabeled
Format Lyophilized
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87676 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87676
100 µg
Each for $770.50
Add to cart
 
Description

Description

CD96 is a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gamma-delta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Lyophilized
Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Nucleocapsid; SARS-CoV2; SARS-CoV-2
RUO
Virus
SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
Unlabeled
PBS with no preservative
HEK293 cells
-20°Cor to +80°C if preferred
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit