Learn More
BioworldSupplier Diversity Partner HUMAN CD99 (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220602821
Recombinant Human CD99 Source: E.Coli Background: CD99 is an integral membrane protein of 185 amino acids that is expressed highly on cells of the hematopoietic system and on Ewing tumor cells. CD99 has been implicated in cell adhesi...
Catalog No. 50-253-381