Learn More
Abnova™ Human CDC40 Full-length ORF (NP_056975.1, 1 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_056975.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51362 |
Molecular Weight (g/mol) | 91.9kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-938-891
|
Abnova™
H00051362P01L |
25 ug |
Each for $769.00
|
|
|||||
89-938-890
|
Abnova™
H00051362P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is found to be essential for the catalytic step II in pre-mRNA splicing process. It is found in the spliceosome, and contains seven WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp17 protein, which functions in two different cellular processes: pre-mRNA splicing and cell cycle progression. It suggests that this protein may play a role in cell cycle progression. [provided by RefSeq]
Sequence: MSAAIAALAASYGSGSGSESDSDSESSRCPLPAADSLMHLTKSPSSKPSLAVAVDSAPEVAVKEDLETGVHLDPAVKEVQYNPTYETMFAPEFGPENPFRTQQMAAPRNMLSGYAEPAHINDFMFEQQRRTFATYGYALDPSLDNHQVSAKYIGSVEEAEKNQGLTVFETGQKKTEKRKKFKENDASNIDGFLGPWAKYVDEKDVAKPSEEEQKELDEITAKRQKKGKQEEEKPGEEKTILHVKEMYDYQGRSYLHIPQDVGVNLRSTMPPEKCYLPKKQIHVWSGHTKGVSAVRLFPLSGHLLLSCSMDCKIKLWEVYGERRCLRTFIGHSKAVRDICFNTAGTQFLSAAYDRYLKLWDTETGQCISRFTNRKVPYCVKFNPDEDKQNLFVAGMSDKKIVQWDIRSGEIVQEYDRHLGAVNTIVFVDENRRFVSTSDDKSLRVWEWDIPVDFKYIAEPSMHSMPAVTLSPNGKWLACQSMDNQILIFGAQNRFRLNKKKIFKGHMVAGYACQVDFSPDMSYVISGDGNGKLNIWDWKTTKLYSRFKAHDKVCIGAVWHPHETSKVITCGWDGLIKLWDSpecifications
NP_056975.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSAAIAALAASYGSGSGSESDSDSESSRCPLPAADSLMHLTKSPSSKPSLAVAVDSAPEVAVKEDLETGVHLDPAVKEVQYNPTYETMFAPEFGPENPFRTQQMAAPRNMLSGYAEPAHINDFMFEQQRRTFATYGYALDPSLDNHQVSAKYIGSVEEAEKNQGLTVFETGQKKTEKRKKFKENDASNIDGFLGPWAKYVDEKDVAKPSEEEQKELDEITAKRQKKGKQEEEKPGEEKTILHVKEMYDYQGRSYLHIPQDVGVNLRSTMPPEKCYLPKKQIHVWSGHTKGVSAVRLFPLSGHLLLSCSMDCKIKLWEVYGERRCLRTFIGHSKAVRDICFNTAGTQFLSAAYDRYLKLWDTETGQCISRFTNRKVPYCVKFNPDEDKQNLFVAGMSDKKIVQWDIRSGEIVQEYDRHLGAVNTIVFVDENRRFVSTSDDKSLRVWEWDIPVDFKYIAEPSMHSMPAVTLSPNGKWLACQSMDNQILIFGAQNRFRLNKKKIFKGHMVAGYACQVDFSPDMSYVISGDGNGKLNIWDWKTTKLYSRFKAHDKVCIGAVWHPHETSKVITCGWDGLIKLWD | |
RUO | |
CDC40 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51362 | |
CDC40 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EHB3/FLJ10564/MGC102802/PRP17/PRPF17 | |
CDC40 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.