Learn More
Novus Biologicals™ Human CDK4 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275171
Catalog No. NBP275171
Add to cart
Description
Description
The Human CDK4 ELISA Kit (Colorimetric) measures human CDK4 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CDK4 in samples. No significant cross-reactivity or interference between Human CDK4 and analogues was observed.
1019
0.19 ng/mL
CDK4
CDK4
Biological Fluids, Plasma, Serum