Learn More
Abnova™ Human CDK5 (NM_004935|NM_003885) Full-length Recombinant Protein with GST-tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3851
Catalog No. 89-939-036
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Insect cell (Sf9) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
1020
CDK5 (Human) Recombinant Protein
>90% by densitometry
5 ug
RUO
CDK5
The specific activity was determined to be 686nmol /min/mg as per Activity Assay Protocol.
CDK5 Approx. MW 59 and p25 Approx. MW 49.
Yes
NM_004935;NM_003885
In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol)
59, 49kDa
Insect cell (Sf9) expression system
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PSSALRE
CDK5
CDK5 Approx. MW 59 and p25 Approx. MW 49.
Insect
GST