Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CDKL3 Partial ORF (AAH41799, 483 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051265Q01L

 View more versions of this product

Catalog No. 89-939-075


Add to cart

Description

Description

The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: GPIQSQMEKGIFNERTGHSDQMANENKRKLNFSRSDRKEFHFPELPVTIQSKDTKGMEVKQIKMLKRESKKTESSKIPTLLNVDQNQEKQEGGDGHCEGKNLKRNRFFFW
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GPIQSQMEKGIFNERTGHSDQMANENKRKLNFSRSDRKEFHFPELPVTIQSKDTKGMEVKQIKMLKRESKKTESSKIPTLLNVDQNQEKQEGGDGHCEGKNLKRNRFFFW
RUO
CDKL3
Wheat Germ (in vitro)
GST
AAH41799
Liquid
51265
CDKL3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NKIAMRE
CDKL3
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit