Learn More
Novus Biologicals Human CENPE ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP275228
Description
The Human CENPE ELISA Kit (Colorimetric) measures human CENPE in serum, plasma and other biological fluids.Specifications
0.16 - 10 ng/mL | |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent. | |
Human | |
1 Kit |
This kit recognizes Human CENPE in samples. No significant cross-reactivity or interference between Human CENPE and analogues was observed. | |
1062 | |
0.10 ng/mL | |
CENPE | |
CENPE | |
Biological Fluids, Plasma, Serum |