Learn More
Abnova™ Human CENPT Full-length ORF (NP_079358.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00080152P01
Description
The centromere is a specialized chromatin domain, present throughout the cell cycle, that acts as a platform on which the transient assembly of the kinetochore occurs during mitosis. All active centromeres are characterized by the presence of long arrays of nucleosomes in which CENPA (MIM 117139) replaces histone H3 (see MIM 601128). CENPT is an additional factor required for centromere assembly (Foltz et al., 2006 [PubMed 16622419]).[supplied by OMIM]
Sequence: MADHNPDSDSTPRTLLRRVLDTADPRTPRRPRSARAGARRALLETASPRKLSGQTRTIARGRSHGARSVGRSAHIQASGHLEEQTPRTLLKNILLTAPESSILMPESVVKPVPAPQAVQPSRQESSCGSLELQLPELEPPTTLAPGLLAPGRRKQRLRLSVFQQGVDQGLSLSQEPQGNADASSLTRSLNLTFATPLQPQSVQRPGLARRPPARRAVDVGAFLRDLRDTSLAPPNIVLEDTQPFSQPMVGSPNVYHSLPCTPHTGAEDAEQAAGRKTQSSGPGLQKNSECVALVAWSQISpecifications
NP_079358.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C16orf56/CENP-T/FLJ13111/FLJ43376 | |
CENPT | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80152 | |
CENPT (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADHNPDSDSTPRTLLRRVLDTADPRTPRRPRSARAGARRALLETASPRKLSGQTRTIARGRSHGARSVGRSAHIQASGHLEEQTPRTLLKNILLTAPESSILMPESVVKPVPAPQAVQPSRQESSCGSLELQLPELEPPTTLAPGLLAPGRRKQRLRLSVFQQGVDQGLSLSQEPQGNADASSLTRSLNLTFATPLQPQSVQRPGLARRPPARRAVDVGAFLRDLRDTSLAPPNIVLEDTQPFSQPMVGSPNVYHSLPCTPHTGAEDAEQAAGRKTQSSGPGLQKNSECVALVAWSQI | |
RUO | |
CENPT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.