Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human CEP57 Partial ORF (NP_055494.2, 19 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_055494.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9702 |
Molecular Weight (g/mol) | 37.84kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-939-274
|
Abnova™
H00009702Q02L |
25 ug |
Each for $769.00
|
|
|||||
89-939-273
|
Abnova™
H00009702Q02S |
10 ug |
Each for $499.00
|
|
|||||
Description
Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).[supplied by OMIM]
Sequence: AEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQESpecifications
NP_055494.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0092/PIG8/TSP57 | |
CEP57 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9702 | |
CEP57 (Human) Recombinant Protein (Q02) | |
AEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQE | |
RUO | |
CEP57 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title