Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CES3 Full-length ORF (AAH53670.1, 1 a.a. - 568 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023491P01L

 View more versions of this product

Catalog No. 89-939-294


Add to cart

Description

Description

Carboxylesterase 3 is a member of a large multigene family. The enzymes encoded by these genes are responsible for the hydrolysis of ester- and amide-bond-containing drugs such as cocaine and heroin. They also hydrolize long-chain fatty acid esters and thioesters. The specific function of this enzyme has not yet been determined; however, it is speculated that carboxylesterases may play a role in lipid metabolism and/or the blood-brain barrier system. [provided by RefSeq]

Sequence: MERAVRVESGVLVGVVCLLLACPATATGPEVAQPEVDTTLGRVRGRQVGVKGTDRLVNVFLGIPFAQPPLGPDRFSAPHPAQPWEGVRDASTAPPMCLQDVESMNSSRFVLNGKQQIFSVSEDCLVLNVYSPAEVPAGSGRPVMVWVHGGALITGAATSYDGSALAAYGDVVVVTVQYRLGVLGFFSTGDEHAPGNQGFLDVVAALRWVQENIAPFGGDLNCVTVFGGSAGGSIISGLVLSPVAAGLFHRAITQSGVITTPGIIDSHPWPLAQKIANTLACSSSSPAEMVQCLQQKEGEELVLSKKLKNTIYPLTVDGTVFPKSPKELLKEKPFHSVPFLMGVNNHEFSWLIPRGWGLLDTMEQMSREDMLAISTPVLTSLDVPPEMMPTVIDEYLGSNSDAQAKCQAFQEFMGDVFINVPTVSFSRYLRDSGSPVFFYEFQHRPSSFAKIKPAWVKADHGAEGAFVFGGPFLMDESSRLEATEEEKQLSLTMMAQWTHFARTGDPNSKALPPWPQFNQAEQYLEINPVPRAGQKFREAWMQFWSETLPSKIQQWHQKQKNRKAQEDL
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ES31/FLJ21736
CES3
Yes
AAH53670.1
Liquid
23491
CES3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERAVRVESGVLVGVVCLLLACPATATGPEVAQPEVDTTLGRVRGRQVGVKGTDRLVNVFLGIPFAQPPLGPDRFSAPHPAQPWEGVRDASTAPPMCLQDVESMNSSRFVLNGKQQIFSVSEDCLVLNVYSPAEVPAGSGRPVMVWVHGGALITGAATSYDGSALAAYGDVVVVTVQYRLGVLGFFSTGDEHAPGNQGFLDVVAALRWVQENIAPFGGDLNCVTVFGGSAGGSIISGLVLSPVAAGLFHRAITQSGVITTPGIIDSHPWPLAQKIANTLACSSSSPAEMVQCLQQKEGEELVLSKKLKNTIYPLTVDGTVFPKSPKELLKEKPFHSVPFLMGVNNHEFSWLIPRGWGLLDTMEQMSREDMLAISTPVLTSLDVPPEMMPTVIDEYLGSNSDAQAKCQAFQEFMGDVFINVPTVSFSRYLRDSGSPVFFYEFQHRPSSFAKIKPAWVKADHGAEGAFVFGGPFLMDESSRLEATEEEKQLSLTMMAQWTHFARTGDPNSKALPPWPQFNQAEQYLEINPVPRAGQKFREAWMQFWSETLPSKIQQWHQKQKNRKAQEDL
RUO
CES3
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit