Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CFC1 Partial ORF (NP_115934.1, 27 a.a. - 93 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name CFC1 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 33.11kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-939-313
View Documents
Abnova™
H00055997Q01L
25 ug
Each for $709.50
Add to cart
 
89-939-312
View Documents
Abnova™
H00055997Q01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene encodes a member of the epidermal growth factor (EGF)- Cripto, Frl-1, and Cryptic (CFC) family. EGF-CFC family member proteins share a variant EGF-like motif, a conserved cysteine-rich domain, and a C-terminal hydrophobic region. These proteins play key roles in intercellular signaling pathways during vertebrate embryogenesis. Mutations in this gene can cause autosomal visceral heterotaxy. This protein is involved in left-right asymmetric morphogenesis during organ development. [provided by RefSeq]

Sequence: QREKHNGGREEVTKVATQKHRQSPLNWTSSHFGEVTGSAEGWGPEEPLPYSRAFGEGASARPRCCRN
Specifications

Specifications

CFC1 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
33.11kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit