Learn More
Abnova™ Human CHAF1A Partial ORF (AAH67093, 21 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010036Q01S
Description
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B; MIM 601245), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro (Kaufman et al., 1995 [PubMed 7600578]).[supplied by OMIM]
Sequence: CKDRPAFPVKKLIQARLPFKRLNLVPKGKADDMSDDQGTSVQSKSPDLEASLDTLENNCHVGSDIDFRPKLVNGKGPLDNFLRNRIETSIGQSTVIIDLTSpecifications
AAH67093 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CKDRPAFPVKKLIQARLPFKRLNLVPKGKADDMSDDQGTSVQSKSPDLEASLDTLENNCHVGSDIDFRPKLVNGKGPLDNFLRNRIETSIGQSTVIIDLT | |
RUO | |
CHAF1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10036 | |
CHAF1A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAF-1/CAF1/CAF1B/CAF1P150/MGC71229/P150 | |
CHAF1A | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.