Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human CHAF1A Partial ORF (AAH67093, 21 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$487.50 - $752.00
Specifications
Accession Number | AAH67093 |
---|---|
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Format | Liquid |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10036 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-939-406
|
Abnova™
H00010036Q01L |
25 ug |
Each for $752.00
|
|
89-939-405
|
Abnova™
H00010036Q01S |
10 ug |
Each for $487.50
|
|
Description
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B; MIM 601245), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro (Kaufman et al., 1995 [PubMed 7600578]).[supplied by OMIM]
Sequence: CKDRPAFPVKKLIQARLPFKRLNLVPKGKADDMSDDQGTSVQSKSPDLEASLDTLENNCHVGSDIDFRPKLVNGKGPLDNFLRNRIETSIGQSTVIIDLTSpecifications
AAH67093 | |
Liquid | |
10036 | |
CHAF1A (Human) Recombinant Protein (Q01) | |
CKDRPAFPVKKLIQARLPFKRLNLVPKGKADDMSDDQGTSVQSKSPDLEASLDTLENNCHVGSDIDFRPKLVNGKGPLDNFLRNRIETSIGQSTVIIDLT | |
RUO | |
CHAF1A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAF-1/CAF1/CAF1B/CAF1P150/MGC71229/P150 | |
CHAF1A | |
Yes | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title