Learn More
Novus Biologicals™ Human CHD3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275266
Catalog No. NBP275266
Add to cart
Description
Description
The Human CHD3 ELISA Kit (Colorimetric) measures human CHD3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human CHD3 in samples. No significant cross-reactivity or interference between Human CHD3 and analogues was observed.
1107
0.19 ng/mL
CHD3
CHD3
Biological Fluids, Plasma, Serum