Learn More
Raybiotech IncSupplier Diversity Partner Human CHI3L1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHCHI3L11
For cell culture supernatants, plasma, and serum samples. Sensitivity: 6 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human CHI3L1. Other species not determined.
Catalog No. 50-194-9118