Learn More
Novus Biologicals™ Human Chromogranin A ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP267242
Catalog No. NBP267242
Add to cart
Description
Description
The Human Chromogranin A ELISA Kit (Colorimetric) measures human Chromogranin A in serum, plasma and other biological fluids.
Specifications
Specifications
62.50 - 4000 pg/mL
This kit recognizes Human CHGA in samples. No significant cross-reactivity or interference between Human CHGA and analogues was observed.
1113
Human
1 Kit
Storage is content dependent.
37.50 pg/mL
ELISA
CHGA
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
Chromogranin A