Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CHST11 Partial ORF (NP_060883, 230 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name CHST11 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 37.62kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-939-578
View Documents
Abnova™
H00050515Q01L
25 ug
Each for $709.50
Add to cart
 
89-939-577
View Documents
Abnova™
H00050515Q01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

Chondroitin 4-sulfotransferases, such as CHST11, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate to position 4 of N-acetylgalactosamine residues in chondroitin (Yamauchi et al., 2000 [PubMed 10722746]).[supplied by OMIM]

Sequence: RKGDDVKFEEFVAYLIDPHTQREEPFNEHWQTVYSLCHPCHIHYDLVGKYETLEEDSNYVLQLAGVGSYLKFPTYAKSTRTTDEMTTEFFQNISSEHQTQLYEVYKLD
Specifications

Specifications

CHST11 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
37.62kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit