Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CIDEC Partial ORF (NP_071377.2, 53 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00063924Q01S

 View more versions of this product

Catalog No. 89-939-638


Add to Cart

Description

Description

DNA fragmentation factor (DFF) induces the fragmentation of DNA associated with apoptosis. A novel family of cell death-inducing DFF45 (MIM 601882)-like effectors (CIDEs), including CIDEC, can also promote apoptosis (Liang et al., 2003 [PubMed 12429024]).[supplied by OMIM]

Sequence: SVRKGIMAYSLEDLLLKVRDTLMLADKPFFLVLEEDGTTVETEEYFQALAGDTVFMVLQKGQKWQPPSEQGTRHPLSLSHKPAKKIDVA
Specifications

Specifications

NP_071377.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SVRKGIMAYSLEDLLLKVRDTLMLADKPFFLVLEEDGTTVETEEYFQALAGDTVFMVLQKGQKWQPPSEQGTRHPLSLSHKPAKKIDVA
RUO
CIDEC
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
63924
CIDEC (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CIDE-3/FLJ20871/Fsp27
CIDEC
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit