Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CKMT1B Full-length ORF (NP_066270.1, 1 a.a. - 417 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001159P01L

 View more versions of this product

Catalog No. 89-939-693


Add to cart

Description

Description

Mitochondrial creatine (MtCK) kinase is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Many malignant cancers with poor prognosis have shown overexpression of ubiquitous mitochondrial creatine kinase; this may be related to high energy turnover and failure to eliminate cancer cells via apoptosis. Ubiquitous mitochondrial creatine kinase has 80% homology with the coding exons of sarcomeric mitochondrial creatine kinase. Two genes located near each other on chromosome 15 have been identified which encode identical mitochondrial creatine kinase proteins. [provided by RefSeq]

Sequence: MAGPFSRLLSARPGLRLLALAGAGSLAAGFLLRPEPVRAASERRRLYPPSAEYPDLRKHNNCMASHLTPAVYARLCDKTTPTGWTLDQCIQTGVDNPGHPFIKTVGMVAGDEETYEVFADLFDPVIQERHNGYDPRTMKHTTDLDASKIRSGYFDERYVLSSRVRTGRSIRGLSLPPACTRAERREVERVVVDALSGLKGDLAGRYYRLSEMTEAEQQQLIDDHFLFDKPVSPLLTAAGMARDWPDARGIWHNNEKSFLIWVNEEDHTRVISMEKGGNMKRVFERFCRGLKEVERLIQERGWEFMWNERLGYILTCPSNLGTGLRAGVHIKLPLLSKDSRFPKILENLRLQKRGTGGVDTAATGGVFDISNLDRLGKSEVELVQLVIDGVNYLIDCERRLERGQDIRIPTPVIHTKH
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CKMT/CKMT1/UMTCK
CKMT1B
Yes
NP_066270.1
Liquid
1159
CKMT1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGPFSRLLSARPGLRLLALAGAGSLAAGFLLRPEPVRAASERRRLYPPSAEYPDLRKHNNCMASHLTPAVYARLCDKTTPTGWTLDQCIQTGVDNPGHPFIKTVGMVAGDEETYEVFADLFDPVIQERHNGYDPRTMKHTTDLDASKIRSGYFDERYVLSSRVRTGRSIRGLSLPPACTRAERREVERVVVDALSGLKGDLAGRYYRLSEMTEAEQQQLIDDHFLFDKPVSPLLTAAGMARDWPDARGIWHNNEKSFLIWVNEEDHTRVISMEKGGNMKRVFERFCRGLKEVERLIQERGWEFMWNERLGYILTCPSNLGTGLRAGVHIKLPLLSKDSRFPKILENLRLQKRGTGGVDTAATGGVFDISNLDRLGKSEVELVQLVIDGVNYLIDCERRLERGQDIRIPTPVIHTKH
RUO
CKMT1B
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit