Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Novus Biologicals™ Human CLEC10A/CD301 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Product Type Recombinant Protein Antigen
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol CLEC10A
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB59659250U View Documents Novus Biologicals™
NBP259659250UG
250μg Each for $1,025.00
N/A
NB5965950UG View Documents Novus Biologicals™
NBP25965950UG
50μg Each for $287.00
N/A
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 61-292 of Human CLEC10A/CD301 The Recombinant Human CLEC10A/CD301 Protein is derived from Baculovirus. The Recombinant Human CLEC10A/CD301 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Recombinant Protein Antigen
RUO
CLEC10A
CLEC10A Recombinant Protein Antigen
In PBS (pH 7.4)
Baculovirus
Unlabeled
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
CD301 antigen, CD301, CD301C-type lectin domain family 10 member A, CLEC10A
>90% by SDS-PAGE
Human
PAGE

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit