Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Clnk Full-Length Orf (Bag52635.1, 1 A.A.-428 A.A.) Recombinant Protein With Gst Tag At N-Terminal.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

Manufacturer:  Abnova™ H00116449P01L

Catalog No. 89-032-137


Add to cart

Description

Description

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: MNRQGNRKTTKEGSNDLKFQNFSLPKNRSWPRINSATGQYQRMNKPLLDWERNFAAVLDGAKGHSDDDYDDPELRMEETWQSIKILPARPIKESEYADTHYFKVAMDTPLPLDTRTSISIGQPTWNTQTRLERVDKPISKDVRSQN

Specifications

Specifications

BAG52635.1
Liquid
116449
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC35197|MIST
CLNK
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
CLNK (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MNRQGNRKTTKEGSNDLKFQNFSLPKNRSWPRINSATGQYQRMNKPLLDWERNFAAVLDGAKGHSDDDYDDPELRMEETWQSIKILPARPIKESEYADTHYFKVAMDTPLPLDTRTSISIGQPTWNTQTRLERVDKPISKDVRSQNIKGDASVRKNKIPLPPPRPLITLPKKYQPLPPEPESSRPPLSQRHTFPEVQRMPSQISLRDLSEVLEAEKVPHNQRKPESTHLLENQNTQEIPLAISSSSFTTSNHSVQNRDHRGGMQPCSPQRCQPPASCSPHENILPYKYTSWRPPFPKRSDRKDVQHNEWYIGEYSRQAVEEAFMKENKDGSFLVRDCSTKSKEEPYVLAVFYENKVYNVKIRFLERNQQFALGTGLRGDEKFDSVEDIIEHYKNFPIILIDGKDKTGVHRKQCHLTQPLPLTRHLLPL
RUO
CLNK
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only