Learn More
Novus Biologicals™ Human CLS1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275339
Description
The Human CLS1 ELISA Kit (Colorimetric) measures human CLS1 in serum, plasma and other biological fluids.Specifications
1.56 - 100 ng/mL | |
This kit recognizes Human CLS in samples. No significant cross-reactivity or interference between Human CLS and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
54675 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
0.94 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CRLS1 | |
1 Kit | |
CLS1 |