Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CNO Full-length ORF (AAH67815.1, 1 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00055330P01L

 View more versions of this product

Catalog No. 89-939-996


Add to Cart

Description

Description

This intronless gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and is a model for Hermansky-Pudlak syndrome. The encoded protein may play a role in intracellular vesicular trafficking. [provided by RefSeq]

Sequence: MEGSFSDGGALPEGLAEEAEPQGAAWSGDSGTVSQSHSSASGPWEDEGAEDGAPGRDLPLHRRAAAGYAACLLPGAGARPEVEALDASLEDLLTRVDEFVGMLDMLRGDSSHVVSEGVPRIHAKAAEMRRIYSRIDRLEAFVRMVGGRVARMEEQVTKAEAELGTFPRAFKKLLHTMNVPSLFSKSAPSRPQQAGYEAPVLFRTEDYFPCCSERPQL
Specifications

Specifications

AAH67815.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCAS4L/FLJ11230
CNO
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55330
CNO (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEGSFSDGGALPEGLAEEAEPQGAAWSGDSGTVSQSHSSASGPWEDEGAEDGAPGRDLPLHRRAAAGYAACLLPGAGARPEVEALDASLEDLLTRVDEFVGMLDMLRGDSSHVVSEGVPRIHAKAAEMRRIYSRIDRLEAFVRMVGGRVARMEEQVTKAEAELGTFPRAFKKLLHTMNVPSLFSKSAPSRPQQAGYEAPVLFRTEDYFPCCSERPQL
RUO
CNO
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit